Assay Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146883.2 | XP_015002369.1 | 753 | 63 | ||
XM_028826274.1 | XP_028682107.1 | 949 | 63 | |||
XM_028826275.1 | XP_028682108.1 | 827 | 63 | |||
XM_028826276.1 | XP_028682109.1 | 813 | 63 | |||
GenBank mRNA | JU333390.1 | - | 703 | 63 | ||
JV460241.1 | - | 750 | 63 | |||
JV528979.1 | - | 755 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
