Assay Details
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103345.4 | XP_001103345.2 | 346 | 61 | ||
XM_002801254.3 | XP_002801300.2 | 353 | 61 | |||
GenBank mRNA | JV658506.1 | - | 1 - 2 | 150 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
