Assay Details
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266557.1 | NP_001253486.1 | 817 | 68 | ||
GenBank mRNA | JU333908.1 | - | 817 | 68 | ||
JU475095.1 | - | 817 | 68 | |||
JV046705.1 | - | 675 | 68 | |||
JV647834.1 | - | 820 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
