Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 15 - 16 | 2358 | 135 | |
XM_028844953.1 | XP_028700786.1 | 2353 | 135 | |||
XM_028844955.1 | XP_028700788.1 | 2326 | 135 | |||
GenBank mRNA | JU321423.1 | - | 15 - 16 | 2296 | 135 | |
JV709601.1 | - | 15 - 16 | 2116 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
