Assay Details
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148410.2 | XP_015003896.1 | 1303 | 76 | ||
XM_015148412.2 | XP_015003898.1 | 1171 | 76 | |||
XM_028826981.1 | XP_028682814.1 | 2308 | 76 | |||
GenBank mRNA | JU335112.1 | - | 2 - 3 | 1047 | 76 | |
JU470643.1 | - | 2 - 3 | 1053 | 76 | ||
JV047453.1 | - | 2 - 3 | 1054 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
