Assay Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194610.2 | NP_001181539.1 | 2 - 3 | 407 | 121 | |
GenBank mRNA | JU480493.1 | - | 2 - 3 | 321 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
