Assay Details
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084312.4 | XP_001084312.2 | 451 | 88 | ||
XM_015141347.2 | XP_014996833.2 | 339 | 88 | |||
XM_028849887.1 | XP_028705720.1 | 2332 | 88 | |||
XM_028849888.1 | XP_028705721.1 | 452 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
