Assay Details
Gene Symbol: SS18L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148654.2 | XP_015004140.2 | 835 | 144 | ||
XM_015148655.2 | XP_015004141.2 | 835 | 144 | |||
XM_015148656.2 | XP_015004142.1 | 1061 | 144 | |||
XM_015148657.2 | XP_015004143.1 | 741 | 144 | |||
XM_028828557.1 | XP_028684390.1 | 1389 | 144 | |||
XM_028828558.1 | XP_028684391.1 | 735 | 144 | |||
GenBank mRNA | JU333909.1 | - | 783 | 144 | ||
JU475097.1 | - | 788 | 144 | |||
JV046706.1 | - | 789 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
