Assay Details
Gene Symbol: CLVS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265738.1 | NP_001252667.1 | 748 | 76 | ||
GenBank mRNA | JV564997.1 | - | 748 | 76 |
Gene Symbol: ASPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU334587.1 | - | 6977 | 76 | ||
JV715524.1 | - | 7006 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
