Assay Details
Gene Symbol: PSMB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136086.2 | XP_014991572.2 | 813 | 85 | ||
XM_015136087.2 | XP_014991573.2 | 946 | 85 | |||
GenBank mRNA | JU335544.1 | - | 5 - 6 | 760 | 85 | |
JU476468.1 | - | 5 - 6 | 783 | 85 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
