Assay Details
Gene Symbol: ST18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145346.2 | XP_015000832.2 | 3532 | 69 | ||
XM_015145347.2 | XP_015000833.2 | 3376 | 69 | |||
XM_015145349.2 | XP_015000835.2 | 3447 | 69 | |||
XM_015145350.2 | XP_015000836.2 | 3453 | 69 | |||
XM_015145351.2 | XP_015000837.2 | 3473 | 69 | |||
XM_028852665.1 | XP_028708498.1 | 3338 | 69 | |||
XM_028852666.1 | XP_028708499.1 | 3291 | 69 | |||
XM_028852667.1 | XP_028708500.1 | 3451 | 69 | |||
XM_028852668.1 | XP_028708501.1 | 3297 | 69 | |||
XM_028852669.1 | XP_028708502.1 | 3368 | 69 | |||
XM_028852670.1 | XP_028708503.1 | 3372 | 69 | |||
XM_028852671.1 | XP_028708504.1 | 3212 | 69 | |||
GenBank mRNA | JU337284.1 | - | 2704 | 69 | ||
JU337285.1 | - | 2704 | 69 | |||
JV727697.1 | - | 2660 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
