Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028851209.1 | XP_028707042.1 | 3001 | 95 | ||
GenBank mRNA | JU479652.1 | - | 1352 | 95 | ||
JV641428.1 | - | 1108 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
