Assay Details
Gene Symbol: FOXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261043.1 | NP_001247972.1 | 540 | 110 | ||
XM_015112307.2 | XP_014967793.1 | 729 | 110 | |||
XM_015112309.2 | XP_014967795.1 | 863 | 110 | |||
XM_015112310.2 | XP_014967796.1 | 629 | 110 | |||
XM_015112311.2 | XP_014967797.1 | 627 | 110 | |||
XM_028831621.1 | XP_028687454.1 | 906 | 110 | |||
XM_028831622.1 | XP_028687455.1 | 1088 | 110 | |||
XM_028831623.1 | XP_028687456.1 | 1101 | 110 | |||
XM_028831624.1 | XP_028687457.1 | 612 | 110 | |||
GenBank mRNA | JU334307.1 | - | 629 | 110 | ||
JU334308.1 | - | 629 | 110 | |||
JU475413.1 | - | 540 | 110 | |||
JU475414.1 | - | 540 | 110 |

More Information
Set Membership: |
![]() ![]() |
