Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099152.4 | XP_001099152.1 | 921 | 84 | ||
XM_028852638.1 | XP_028708471.1 | 296 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028852638.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
