Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193775.2 | NP_001180704.1 | 1 - 2 | 174 | 73 | |
XM_015138139.2 | XP_014993625.1 | 207 | 73 | |||
GenBank mRNA | JU477127.1 | - | 1 - 2 | 184 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
