Assay Details
Gene Symbol: PNKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028831102.1 | XP_028686935.1 | 290 | 61 | ||
GenBank mRNA | JU327047.1 | - | 1 - 2 | 155 | 61 | |
JV463925.1 | - | 2 - 3 | 193 | 61 | ||
JV540369.1 | - | 2 - 3 | 233 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
