Assay Details
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194143.2 | NP_001181072.1 | 3 - 4 | 897 | 84 | |
GenBank mRNA | JU472964.1 | - | 3 - 4 | 276 | 84 | |
JV632821.1 | - | 3 - 4 | 279 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
