Assay Details
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261519.1 | NP_001248448.1 | 1380 | 99 | ||
XM_015142940.2 | XP_014998426.1 | 1445 | 99 | |||
XM_028850235.1 | XP_028706068.1 | 1431 | 99 | |||
XM_028850236.1 | XP_028706069.1 | 1486 | 99 | |||
XM_028850238.1 | XP_028706071.1 | 1324 | 99 | |||
XM_028850239.1 | XP_028706072.1 | 1301 | 99 | |||
GenBank mRNA | JU320193.1 | - | 1338 | 99 | ||
JV043786.1 | - | 1380 | 99 | |||
JV635706.1 | - | 1387 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
