Assay Details
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265726.1 | NP_001252655.1 | 1 - 2 | 563 | 75 | |
GenBank mRNA | JU326335.1 | - | 1 - 2 | 381 | 75 | |
JV565012.1 | - | 1 - 2 | 563 | 75 | ||
JV705634.1 | - | 1 - 2 | 232 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
