Assay Details
Gene Symbol: MELTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843543.1 | XP_028699376.1 | 648 | 91 | ||
XM_028843544.1 | XP_028699377.1 | 648 | 91 | |||
GenBank mRNA | JU486829.1 | - | 5 - 6 | 634 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
