Assay Details
Gene Symbol: DYNC2I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116677.2 | XP_014972163.1 | 551 | 68 | ||
XM_028834760.1 | XP_028690593.1 | 634 | 68 | |||
GenBank mRNA | JV632640.1 | - | 2 - 3 | 446 | 68 | |
JV664827.1 | - | 2 - 3 | 434 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
