Assay Details
Gene Symbol: CEMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117889.2 | XP_014973375.1 | 4487 | 107 | ||
XM_015117890.2 | XP_014973376.1 | 4054 | 107 | |||
GenBank mRNA | JU470675.1 | - | 4269 | 107 | ||
JV046626.1 | - | 4212 | 107 | |||
JV635496.1 | - | 4259 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
