Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260728.1 | NP_001247657.1 | 298 | 146 | ||
GenBank mRNA | JU320977.1 | - | 281 | 146 | ||
JU472591.1 | - | 279 | 146 | |||
JV045162.1 | - | 298 | 146 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
