Assay Details
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266124.1 | NP_001253053.1 | 321 | 114 | ||
GenBank mRNA | AF252388.1 | - | 118 | 114 | ||
JU326959.1 | - | 122 | 114 | |||
JU473867.1 | - | 298 | 114 | |||
JV045638.1 | - | 321 | 114 | |||
JV637471.1 | - | 314 | 114 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
