Assay Details
Gene Symbol: LZIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149353.2 | XP_015004839.1 | 471 | 131 | ||
XM_015149402.2 | XP_015004888.1 | 471 | 131 | |||
GenBank mRNA | JU336714.1 | - | 391 | 131 | ||
JU471925.1 | - | 383 | 131 | |||
JV048446.1 | - | 366 | 131 | |||
JV663401.1 | - | 363 | 131 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
