Assay Details
Gene Symbol: LOC716834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114328.4 | XP_001114328.2 | 3503 | 64 | ||
XM_015151038.2 | XP_015006524.2 | 3466 | 64 | |||
GenBank mRNA | JU323357.1 | - | 25 - 26 | 3212 | 64 | |
JU471944.1 | - | 25 - 26 | 3193 | 64 | ||
JV044860.1 | - | 25 - 26 | 3223 | 64 | ||
JV452949.1 | - | 25 - 26 | 3236 | 64 | ||
JV635650.1 | - | 25 - 26 | 3190 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
