Assay Details
Gene Symbol: PPP1R14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137129.2 | XP_014992615.1 | 1069 | 71 | ||
GenBank mRNA | JU328496.1 | - | 271 | 71 | ||
JV072533.1 | - | 270 | 71 | |||
JV600321.1 | - | 567 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
