Assay Details
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132817.2 | XP_014988303.1 | 1542 | 73 | ||
XM_028845303.1 | XP_028701136.1 | 1542 | 73 | |||
XM_028845304.1 | XP_028701137.1 | 1950 | 73 | |||
XM_028845305.1 | XP_028701138.1 | 1860 | 73 | |||
XM_028845306.1 | XP_028701139.1 | 1406 | 73 | |||
XM_028845307.1 | XP_028701140.1 | 1542 | 73 | |||
XM_028845308.1 | XP_028701141.1 | 1542 | 73 | |||
GenBank mRNA | JU470963.1 | - | 12 - 13 | 1273 | 73 |
Gene Symbol: LOC114669760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842588.1 | XP_028698421.1 | 1397 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
