Assay Details
Gene Symbol: PAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257252.1 | NP_001244181.1 | 154 | 148 | ||
XM_015140811.2 | XP_014996297.1 | 331 | 148 | |||
GenBank mRNA | CN642792.1 | - | 253 | 148 | ||
JU334834.1 | - | 172 | 148 | |||
JV047268.1 | - | 250 | 148 | |||
JV566125.1 | - | 293 | 148 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
