Assay Details
Gene Symbol: GUCA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085421.4 | XP_001085421.1 | 274 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001085421.4 transcripts.
Set Membership: |
![]() ![]() |
