Assay Details
Gene Symbol: NRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105167.1 | NP_001098637.1 | 500 | 70 | ||
GenBank mRNA | JU473146.1 | - | 509 | 70 | ||
JV641270.1 | - | 493 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
