Assay Details
Gene Symbol: COASY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109889.4 | XP_001109889.3 | 1597 | 91 | ||
XM_015119715.2 | XP_014975201.2 | 2171 | 91 | |||
XM_015119716.2 | XP_014975202.2 | 1783 | 91 | |||
GenBank mRNA | JV594645.1 | - | 4 - 5 | 1714 | 91 | |
JV662280.1 | - | 4 - 5 | 1718 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
