Assay Details
Gene Symbol: NREP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193355.3 | NP_001180284.1 | 58 | 78 | ||
XM_015140564.2 | XP_014996050.1 | 265 | 78 | |||
XM_015140565.2 | XP_014996051.1 | 368 | 78 | |||
XM_028849263.1 | XP_028705096.1 | 236 | 78 | |||
XM_028849264.1 | XP_028705097.1 | 273 | 78 | |||
XM_028849265.1 | XP_028705098.1 | 246 | 78 | |||
GenBank mRNA | JU322948.1 | - | 171 | 78 | ||
JV636844.1 | - | 172 | 78 | |||
JV636845.1 | - | 175 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193355.3, XM_015140564.2, XM_015140565.2, XM_028849263.1, XM_028849264.1, XM_028849265.1 transcripts.
Set Membership: |
![]() ![]() |
