Assay Details
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093274.4 | XP_001093274.2 | 545 | 98 | ||
XM_015142682.2 | XP_014998168.2 | 545 | 98 | |||
XM_028851103.1 | XP_028706936.1 | 721 | 98 | |||
GenBank mRNA | JU328507.1 | - | 412 | 98 | ||
JU473028.1 | - | 501 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
