Assay Details
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145024.2 | XP_015000510.2 | 1145 | 67 | ||
XM_015145025.2 | XP_015000511.2 | 1027 | 67 | |||
XM_015145026.2 | XP_015000512.2 | 967 | 67 | |||
XM_028852483.1 | XP_028708316.1 | 4716 | 67 | |||
XM_028852484.1 | XP_028708317.1 | 1186 | 67 | |||
XM_028852485.1 | XP_028708318.1 | 4598 | 67 | |||
XM_028852486.1 | XP_028708319.1 | 951 | 67 | |||
XM_028852487.1 | XP_028708320.1 | 1144 | 67 | |||
XM_028852488.1 | XP_028708321.1 | 4597 | 67 | |||
XM_028852489.1 | XP_028708322.1 | 966 | 67 | |||
XM_028852490.1 | XP_028708323.1 | 929 | 67 | |||
XM_028852491.1 | XP_028708324.1 | 1099 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028852490.1, XM_028852491.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
