Assay Details
Gene Symbol: GCLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145343.2 | XP_015000829.1 | 964 | 102 | ||
GenBank mRNA | JU334027.1 | - | 5 - 6 | 541 | 102 | |
JU475202.1 | - | 5 - 6 | 651 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
