Assay Details
Gene Symbol: SLC31A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117052.2 | XP_014972538.1 | 389 | 65 | ||
XM_028834965.1 | XP_028690798.1 | 353 | 65 | |||
GenBank mRNA | JU322897.1 | - | 2 - 3 | 158 | 65 | |
JU472705.1 | - | 2 - 3 | 175 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
