Assay Details
Gene Symbol: ELK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194388.2 | NP_001181317.1 | 4 - 5 | 1239 | 77 | |
XM_015152478.2 | XP_015007964.1 | 1454 | 77 | |||
XM_028828962.1 | XP_028684795.1 | 1146 | 77 | |||
GenBank mRNA | JU471176.1 | - | 4 - 5 | 1387 | 77 | |
JV046394.1 | - | 5 - 6 | 1276 | 77 | ||
JV636013.1 | - | 4 - 5 | 1305 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
