Assay Details
Gene Symbol: BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146598.2 | XP_015002084.1 | 659 | 74 | ||
XM_015146599.2 | XP_015002085.1 | 652 | 74 | |||
XM_015146600.2 | XP_015002086.1 | 431 | 74 | |||
XM_015146601.2 | XP_015002087.1 | 357 | 74 | |||
XM_015146602.2 | XP_015002088.1 | 421 | 74 | |||
GenBank mRNA | JV626022.1 | - | 891 | 74 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
