Assay Details
Gene Symbol: MED26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123227.2 | XP_014978713.1 | 488 | 51 | ||
GenBank mRNA | JU477904.1 | - | 1 - 2 | 151 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() |
