Assay Details
Gene Symbol: TENT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261739.2 | NP_001248668.1 | 1151 | 131 | ||
XM_015140203.2 | XP_014995689.1 | 1249 | 131 | |||
XM_015140204.2 | XP_014995690.1 | 1283 | 131 | |||
XM_015140207.2 | XP_014995693.1 | 1237 | 131 | |||
XM_028849246.1 | XP_028705079.1 | 1184 | 131 | |||
GenBank mRNA | JU321814.1 | - | 1108 | 131 | ||
JU321818.1 | - | 1216 | 131 | |||
JU333207.1 | - | 1204 | 131 | |||
JU471567.1 | - | 1228 | 131 | |||
JU471568.1 | - | 1197 | 131 | |||
JU474524.1 | - | 1185 | 131 | |||
JV046328.1 | - | 1213 | 131 | |||
JV046329.1 | - | 1225 | 131 | |||
JV046330.1 | - | 1152 | 131 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
