Assay Details
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260739.1 | NP_001247668.1 | 3 - 4 | 450 | 68 | |
GenBank mRNA | JU320685.1 | - | 3 - 4 | 436 | 68 | |
JU472487.1 | - | 3 - 4 | 471 | 68 | ||
JV047478.1 | - | 3 - 4 | 450 | 68 | ||
JV654843.1 | - | 3 - 4 | 475 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
