Assay Details
Gene Symbol: TRMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149742.2 | XP_015005228.1 | 1075 | 100 | ||
GenBank mRNA | JU333989.1 | - | 914 | 100 | ||
JU333990.1 | - | 914 | 100 | |||
JU475173.1 | - | 907 | 100 | |||
JU475174.1 | - | 907 | 100 | |||
JV046757.1 | - | 958 | 100 | |||
JV046758.1 | - | 958 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
