Assay Details
Gene Symbol: VAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032820.2 | NP_001027992.1 | 2 - 3 | 223 | 79 | |
GenBank mRNA | AF240769.1 | - | 2 - 3 | 217 | 79 | |
JV044681.1 | - | 2 - 3 | 210 | 79 | ||
JV565238.1 | - | 2 - 3 | 222 | 79 | ||
JV635626.1 | - | 2 - 3 | 225 | 79 | ||
JV669158.1 | - | 2 - 3 | 223 | 79 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
