Assay Details
Gene Symbol: RSKR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108797.4 | XP_001108797.2 | 859 | 91 | ||
XM_015118901.2 | XP_014974387.1 | 856 | 91 | |||
XM_015118902.2 | XP_014974388.1 | 859 | 91 | |||
XM_015118903.2 | XP_014974389.1 | 706 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
