Assay Details
Gene Symbol: RSKR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108797.4 | XP_001108797.2 | 613 | 115 | ||
XM_015118901.2 | XP_014974387.1 | 610 | 115 | |||
XM_015118902.2 | XP_014974388.1 | 613 | 115 |

More Information
Set Membership: |
![]() ![]() |
