Assay Details
Gene Symbol: SAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266332.1 | NP_001253261.1 | 1 - 2 | 533 | 125 | |
GenBank mRNA | JU326394.1 | - | 1 - 2 | 244 | 125 | |
JU473121.1 | - | 1 - 2 | 533 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
