Assay Details
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168328.1 | NP_001161800.1 | 685 | 141 | ||
XM_015141348.2 | XP_014996834.1 | 1355 | 141 | |||
XM_015141350.2 | XP_014996836.1 | 987 | 141 | |||
XM_015141351.2 | XP_014996837.1 | 1077 | 141 | |||
XM_028849100.1 | XP_028704933.1 | 957 | 141 | |||
GenBank mRNA | GU086165.1 | - | 685 | 141 | ||
GU086166.1 | - | 685 | 141 | |||
JU333820.1 | - | 840 | 141 | |||
JV452117.1 | - | 815 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
