Assay Details
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115393.2 | XP_014970879.1 | 1268 | 67 | ||
XM_015115395.2 | XP_014970881.1 | 1268 | 67 | |||
XM_028833900.1 | XP_028689733.1 | 910 | 67 | |||
GenBank mRNA | JU341133.1 | - | 9 - 10 | 1330 | 67 | |
JU341134.1 | - | 9 - 10 | 1330 | 67 | ||
JV565045.1 | - | 8 - 9 | 1168 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
