Assay Details
Gene Symbol: OGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148051.2 | XP_015003537.1 | 2942 | 67 | ||
XM_015148052.2 | XP_015003538.1 | 2783 | 67 | |||
XM_028826799.1 | XP_028682632.1 | 2843 | 67 | |||
GenBank mRNA | JU320291.1 | - | 13 - 14 | 2670 | 67 | |
JU320292.1 | - | 12 - 13 | 2511 | 67 | ||
JU472439.1 | - | 13 - 14 | 2614 | 67 | ||
JV043778.1 | - | 13 - 14 | 2675 | 67 | ||
JV705461.1 | - | 13 - 14 | 2401 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
